CAS RN 690632-70-3
IUPAC Name:
2-bromo-1-(5-bromo-2,3-dihydro-1-benzofuran-7-yl)ethan-1-one
Synonyms:
2-bromo-1-(5-bromo-2,3-dihydro-1-benzofuran-7-yl)ethanone
Molecular Weight (g/mol):
319.98
Molecular Formula:
C10H8Br2O2
InChi Key:
NPGVDFRSCHXENE-UHFFFAOYSA-N
SMILES:
BrCC(=O)C1=CC(Br)=CC2=C1OCC2
1
–
1
of
1
results
1
2-bromo-1-(5-bromo-2,3-dihydro-1-benzofuran-7-yl)ethanone, Thermo Scientific™
CAS: 690632-70-3 Molecular Formula: C10H8Br2O2 Molecular Weight (g/mol): 319.98 MDL Number: MFCD06200889 InChI Key: NPGVDFRSCHXENE-UHFFFAOYSA-N Synonym: 2-bromo-1-5-bromo-2,3-dihydro-1-benzofuran-7-yl ethanone,2-bromo-1-5-bromo-2,3-dihydrobenzofuran-7-yl ethanone,2-bromo-1-5-bromo-2,3-dihydro-1-benzo b furan-7-yl ethanone,2-bromo-1-5-bromo-2,3-dihydro-1-benzofuran-7-yl ethan-1-one PubChem CID: 2794800 IUPAC Name: 2-bromo-1-(5-bromo-2,3-dihydro-1-benzofuran-7-yl)ethanone SMILES: C1COC2=C1C=C(C=C2C(=O)CBr)Br