accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FOOO

CAS RN 69155-76-6

IUPAC Name: 6-fluoro-4-oxo-4H-chromene-3-carbaldehyde
Synonyms: 6-fluoro-4-oxochromene-3-carbaldehyde
Molecular Weight (g/mol): 192.15
Molecular Formula: C10H5FO3
InChi Key: VHRMOTNEBIKURN-UHFFFAOYSA-N
SMILES: FC1=CC=C2OC=C(C=O)C(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results