accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COClOH

CAS RN 6921-66-0

IUPAC Name: 1-(4-chloro-2-hydroxyphenyl)ethan-1-one
Synonyms: 1-(4-chloro-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: QCVSDCHNBNFJDQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(O)C=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)