accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOBr

CAS RN 69240-56-8

IUPAC Name: 4-(4-bromophenoxy)benzaldehyde
Synonyms: 4-(4-bromophenoxy)benzaldehyde
Molecular Weight (g/mol): 277.12
Molecular Formula: C13H9BrO2
InChi Key: WMDDJOBNAOVSLP-UHFFFAOYSA-N
SMILES: BrC1=CC=C(OC2=CC=C(C=O)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results