accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNH2(R)(S)HO

CAS RN 6936-47-6

IUPAC Name: (1S,2R)-2-aminocyclohexan-1-ol hydrochloridyl
Synonyms: (1S,2R)-2-aminocyclohexan-1-ol hydrochloridyl
Molecular Weight (g/mol): 150.63
Molecular Formula: C6H13ClNO
InChi Key: SEBSRZIHRZRNAH-IBTYICNHSA-N
SMILES: [Cl].N[C@@H]1CCCC[C@@H]1O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)