accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 694-72-4

IUPAC Name: octahydropentalene
Synonyms: bicyclo[3.3.0]octane
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: AEBWATHAIVJLTA-UHFFFAOYSA-N
SMILES: C1CC2CCCC2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results