Learn More
Bicyclo[3.3.0]octane, Thermo Scientific™
Product Code. 17525545 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17525545 250 mg 250mg
17515545 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17525545 Supplier Thermo Scientific Acros Supplier No. 468522500
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Chemical Identifiers
CAS 694-72-4
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number 00060882
InChI Key AEBWATHAIVJLTA-UHFFFAOYSA-N
IUPAC Name octahydropentalene
SMILES C1CC2CCCC2C1
Specifications
Alpha Vector BICYCLOOCTANE
Color Colorless
Boiling Point 137°C
Flash Point 26°C
Infrared Spectrum Conforms
Packaging Glass Bottle
Quantity 250 mg
Physical Form Liquid
Chemical Name or Material Bicyclo[3.3.0]octane