accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OF

CAS RN 694-82-6

IUPAC Name: 2-fluorocyclohexan-1-one
Synonyms: 2-fluorocyclohexan-1-one
Molecular Weight (g/mol): 116.14
Molecular Formula: C6H9FO
InChi Key: VQYOFTVCYSPHPG-UHFFFAOYNA-N
SMILES: FC1CCCCC1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results