accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOBrOH3C

CAS RN 6948-30-7

IUPAC Name: 3-bromo-4,5-dimethoxybenzaldehyde
Synonyms: 3-bromo-4,5-dimethoxybenzaldehyde
Molecular Weight (g/mol): 245.07
Molecular Formula: C9H9BrO3
InChi Key: ICVODPFGWCUVJC-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC(Br)=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results