accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 69725-37-7

H3CCH3OOCH3CH3H3C

CAS RN 69725-37-7

IUPAC Name: 2,2,7-trimethyloctane-3,5-dione
Synonyms: 2,2,7-trimethyloctane-3,5-dione
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: KEBPOWLGOOTMPK-UHFFFAOYSA-N
SMILES: CC(C)CC(=O)CC(=O)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)