accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6975-99-1

H3CCH3

CAS RN 6975-99-1

IUPAC Name: dodec-6-yne
Synonyms: dodec-6-yne
Molecular Weight (g/mol): 166.31
Molecular Formula: C12H22
InChi Key: IHPFQAOOSAGSPN-UHFFFAOYSA-N
SMILES: CCCCCC#CCCCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results