Learn More
6-Dodecyne, 98%
Product Code. 11397656
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397656 5 g 5g
11307666 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11397656 Supplier Thermo Scientific Alfa Aesar Supplier No. L11262.06
Request Bulk or Custom Format
CAS: 6975-99-1 | C12H22 | 166.31 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6975-99-1
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00041658
InChI Key IHPFQAOOSAGSPN-UHFFFAOYSA-N
Synonym 6-dodecyne, acmc-209oav
PubChem CID 138890
IUPAC Name dodec-6-yne
SMILES CCCCCC#CCCCCC
Specifications
Density 0.79
Boiling Point 99°C to 100°C (4 mmHg)
Linear Formula CH3(CH2)4C≡C(CH2)4CH3
Refractive Index 1.439
Quantity 5 g
Beilstein 1737397
Formula Weight 166.31
Percent Purity 98%
Chemical Name or Material 6-Dodecyne