accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OClClCH3ONCH3(S)OOHOCH3NH3CCH3(R)OHOH

CAS RN 6989-98-6

IUPAC Name: hydrogen (1S,16R)-9,21-dihydroxy-10,25-dimethoxy-15,15,30-trimethyl-7,23-dioxa-15,30-diazaheptacyclo[22.6.2.2³,⁶.1⁸,¹².1¹⁸,²².0²⁷,³¹.0¹⁶,³⁴]hexatriaconta-3,5,8,10,12(34),18,20,22(33),24(32),25,27(31),35-dodecaen-15-ium pentahydrate dichloride
Synonyms: hydrogen tubocurarine pentahydrate dichloride
Molecular Weight (g/mol): 771.73
Molecular Formula: C37H52Cl2N2O11
InChi Key: WMIZITXEJNQAQK-GGDSLZADSA-N
SMILES: [H+].O.O.O.O.O.[Cl-].[Cl-].COC1=CC2=C3[C@@H](CC4=CC=C(O)C(OC5=CC6=C(CCN(C)[C@H]6CC6=CC=C(OC3=C1O)C=C6)C=C5OC)=C4)[N+](C)(C)CC2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)