Learn More
(+)-Tubocurarine Chloride Hydrochloride Pentahydrate, TRC
Product Code. 30456875
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456875 50 mg 50mg
30454256 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30456875 Supplier Toronto Research Chemicals Supplier No. TRCT94762050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (+)-Tubocurarine chloride hydrochloride pentahydrate
CAS 6989-98-6
Molecular Formula C37 H41 N2 O6 . Cl . Cl H . 5 H2 O
Quantity 50 mg
Synonym 13H-4,6:21,24-Dietheno-8,12-metheno-1H-pyrido[3',2':14,15][1,11]dioxacycloeicosino[2,3,4-ij]isoquinolinium','2,3,13a,14,15,16,25,25a-octahydro-9,19-dihydroxy-18,29-dimethoxy-1,14,14-trimethyl-','chloride','hydrochloride','pentahydrate','(13aR,25aS)-, Tubocuraranium','7',12'-dihydroxy-6,6'-dimethoxy-2,2',2'-trimethyl-','chloride','hydrochloride','pentahydrate, (+)-Tubocurarine chloride hydrochloride pentahydrate, Tubarine pentahydrate, d-Tubocurarine dichloride pentahydrate
InChI Formula InChI=1S/C37H40N2O6.2ClH.5H2O/c1-38-14-12-24-19-32(42-4)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-37-35-25(20-34(43-5)36(37)41)13-15-39(2,3)29(35)17-23-8-11-30(40)31(18-23)45-33;;;;;;;/h6-11,18-21,28-29H,12-17H2,1-5H3,(H-,40,41);2*1H;5*1H2/t28-,29+;;;;;;;/m0......./s1
SMILES O.O.O.O.O.Cl.[Cl-].COc1cc2CC[N+](C)(C)[C@@H]3Cc4ccc(O)c(Oc5cc6[C@H](Cc7ccc(Oc(c1O)c23)cc7)N(C)CCc6cc5OC)c4
IUPAC Name (13aR,25aS)-2,3,13a,14,15,16,25,25a-Octahydro-9,19-dihydroxy-18,29-dimethoxy-1,14,14-trimethyl-13H-4,6:21,24-dietheno-8,12-metheno-1H-pyrido[3',2':14,15][1,11]dioxacycloeicosino[2,3,4-ij]isoquinolinium chloride hydrochloride pentahydrate
Molecular Weight (g/mol) 771.72
Formula Weight 770.2948