accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COFOH

CAS RN 699-92-3

IUPAC Name: 1-(3-fluoro-2-hydroxyphenyl)ethan-1-one
Synonyms: 1-(3-fluoro-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: WXTHZWWWCICGAN-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(O)C(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results