accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 699012-94-7

PtNNNNNaNaHOH

CAS RN 699012-94-7

IUPAC Name: platinum(2+) disodium tetrakis(iminomethanide) hydrate
Synonyms: platinum(2+) disodium tetrakis(cyanide) hydrate
Molecular Weight (g/mol): 363.15
Molecular Formula: C4H2N4Na2OPt
InChi Key: BMZQYSVVRURJND-UHFFFAOYSA-N
SMILES: O.[Na+].[Na+].[Pt++].[C-]#N.[C-]#N.[C-]#N.[C-]#N
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results