accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrO

CAS RN 70-11-1

IUPAC Name: 2-bromo-1-phenylethan-1-one
Synonyms: phenacyl bromide
Molecular Weight (g/mol): 199.05
Molecular Formula: C8H7BrO
InChi Key: LIGACIXOYTUXAW-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)