accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONH2

CAS RN 70-69-9

IUPAC Name: 1-(4-aminophenyl)propan-1-one
Synonyms: 4'-aminopropiophenone
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: FSWXOANXOQPCFF-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results