accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHOO

CAS RN 700-44-7

IUPAC Name: 2-hydroxy-6-methoxybenzaldehyde
Synonyms: 2-hydroxy-6-methoxybenzaldehyde
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: DZJPDDVDKXHRLF-UHFFFAOYSA-N
SMILES: COC1=CC=CC(O)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results