accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOCH3OCH3OH3C

CAS RN 70311-20-5

IUPAC Name: ethyl 3-(3,4,5-trimethoxyphenyl)propanoate
Synonyms: ethyl 3-(3,4,5-trimethoxyphenyl)propanoate
Molecular Weight (g/mol): 268.31
Molecular Formula: C14H20O5
InChi Key: AYBWQZGGKFGQLB-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC1=CC(OC)=C(OC)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results