accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 70356-09-1

IUPAC Name: 1-(4-tert-butylphenyl)-3-(4-methoxyphenyl)propane-1,3-dione
Synonyms: avobenzone avobenzona avobenzone (product) avobenzonum parsol-1789 avobenzone preparation parsol 1789 butyl methoxydibenzoylmethane
Molecular Weight (g/mol): 310.39
Molecular Formula: C20H22O3
InChi Key: XNEFYCZVKIDDMS-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)CC(=O)C1=CC=C(C=C1)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results