missing translation for 'onlineSavingsMsg'
Learn More

Avobenzone, Mikromol

Product Code. 30461582
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30461582 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461582 Supplier Mikromol Supplier No. MM0902.00
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Avobenzone, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Avobenzone
CAS 70356-09-1
Analyte or Component Names Avobenzone
Shipping Condition Room Temperature
Molecular Formula C20 H22 O3
Certifications/Compliance ISO 17034
Synonym 1-[4-(1,1-Dimethylethyl)phenyl]-3-(4-methoxyphenyl)-1,3-propanedione, 1-(4-Methoxyphenyl)-3-(4-tert-butylphenyl)propane-1,3-dione, 1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)propan-1,3-dione, 1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)propane-1,3-dione, 1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)propane-1,3-dione, 1-(4'-tert-Butylphenyl)-3-(4'-methoxyphenyl)propane-1,3-dione, 4-(1,1-Dimethylethyl)-4'-methoxydibenzoylmethane, 4-Methoxy-4'-tert-butyldibenzoylmethane, 4-tert-Butyl-4'-methoxydibenzoylmethane, Avobenzone, Butylmethoxydibenzoylmethane, Escalol 517, Eusolex 9020, NeoHeliopan 357, Parsol 1789, Parsol A, Parsol RTM 1789, Photoplex, Solarom BMBM, Uvinul BMBM
InChI Formula InChI=1S/C20H22O3/c1-20(2,3)16-9-5-14(6-10-16)18(21)13-19(22)15-7-11-17(23-4)12-8-15/h5-12H,13H2,1-4H3
Solution Type Neat
SMILES COc1ccc(cc1)C(=O)CC(=O)c2ccc(cc2)C(C)(C)C
IUPAC Name 1-(4-tert-butylphenyl)-3-(4-methoxyphenyl)propane-1,3-dione
Molecular Weight (g/mol) 310.39
Quantity 500 mg
Formula Weight 310.1569 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.