CAS RN 70476-82-3
IUPAC Name:
dihydrogen 1,4-dihydroxy-5,8-bis({2-[(2-hydroxyethyl)amino]ethyl}amino)-9,10-dihydroanthracene-9,10-dione dichloride
Synonyms:
mitoxantrone
Molecular Weight (g/mol):
517.4
Molecular Formula:
C22H30Cl2N4O6
InChi Key:
ZAHQPTJLOCWVPG-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].OCCNCCNC1=CC=C(NCCNCCO)C2=C1C(=O)C1=C(O)C=CC(O)=C1C2=O
1
–
4
of
4
results
2
Mitoxantrone Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Mitoxantrone Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
Mitoxantrone Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.