accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHH3CO

CAS RN 705-15-7

IUPAC Name: 1-(2-hydroxy-5-methoxyphenyl)ethan-1-one
Synonyms: 1-(2-hydroxy-5-methoxyphenyl)ethanone
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: MLIBGOFSXXWRIY-UHFFFAOYSA-N
SMILES: COC1=CC=C(O)C(=C1)C(C)=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (5)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (3)
  • (5)
  • (2)