accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FBrNOO

CAS RN 7087-65-2

IUPAC Name: 1-bromo-3-fluoro-5-nitrobenzene
Synonyms: 1-bromo-3-fluoro-5-nitrobenzene
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: SWXVEPMSQBEVRH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=CC(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)