accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 70916-98-2

OOHO

CAS RN 70916-98-2

IUPAC Name: 4'-formyl-[1,1'-biphenyl]-4-carboxylic acid
Synonyms: 4'-formyl-[1,1'-biphenyl]-4-carboxylic acid
Molecular Weight (g/mol): 226.23
Molecular Formula: C14H10O3
InChi Key: JCEAFZUEUSBESW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results