accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrCH3NH3CH3CH3C

CAS RN 71-91-0

IUPAC Name: tetraethylazanium bromide
Synonyms: tetraethylammonium bromide tetraethyl ammonium bromide tetrylammonium bromide (inn)
Molecular Weight (g/mol): 210.16
Molecular Formula: C8H20BrN
InChi Key: HWCKGOZZJDHMNC-UHFFFAOYSA-M
SMILES: [Br-].CC[N+](CC)(CC)CC
Molecular Weight (g/mol) 
  • (5)
  • (10)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (6)
  • (4)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (3)
  • (3)
  • (1)
  • (5)
  • (10)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (4)
  • (4)
  • (2)