missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Bromide, for HPLC, Certified, Fisher Chemical

Product Code. 10744001 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10744001 25 g Plastic powder jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10744001 Supplier Fisher Chemical Supplier No. T/0621/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 285°C
CAS 71-91-0
Color Cream
pH 6.5
Physical Form Powder/Solid
Quantity 25 g
Molecular Formula C8H20BrN
MDL Number 11825
Synonym tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
InChI Key HWCKGOZZJDHMNC-UHFFFAOYSA-M
SMILES CC[N+](CC)(CC)CC.[Br-]
IUPAC Name tetraethylazanium;bromide
Molecular Weight (g/mol) 210.159
PubChem CID 6285
Formula Weight 210.16g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Tetraethylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.