accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7143-01-3

H3CSOOOSCH3OO

CAS RN 7143-01-3

IUPAC Name: methanesulfonyl methanesulfonate
Synonyms: methanesulfonic anhydride
Molecular Weight (g/mol): 174.19
Molecular Formula: C2H6O5S2
InChi Key: IZDROVVXIHRYMH-UHFFFAOYSA-N
SMILES: CS(=O)(=O)OS(C)(=O)=O
Boiling Point 
  • (4)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)