missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic Anhydride, TRC

Product Code. 30473475
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473475 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473475 Supplier Toronto Research Chemicals Supplier No. TRCM2262555G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methanesulfonic Anhydride
CAS 7143-01-3
Molecular Formula C2H6O5S2
Quantity 5 g
Synonym Mesyl Anhydride, Methanesulfonyl Anhydride, NSC 65906
InChI Formula InChI=1S/C2H6O5S2/c1-8(3,4)7-9(2,5)6/h1-2H3
SMILES CS(=O)(=O)OS(=O)(=O)C
IUPAC Name methylsulfonyl methanesulfonate
Molecular Weight (g/mol) 174.2
Formula Weight 173.9657
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.