accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CPCH3

CAS RN 71501-08-1

IUPAC Name: dipentyl(phenyl)phosphane
Synonyms: dipentyl(phenyl)phosphane
Molecular Weight (g/mol): 250.37
Molecular Formula: C16H27P
InChi Key: QDZAKCWPRPKCBW-UHFFFAOYSA-N
SMILES: CCCCCP(CCCCC)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results