CAS RN 71510-95-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl-n-methyl malonamide, TRC
CAS: 71510-95-7 Molecular Formula: C6H11NO3 Molecular Weight (g/mol): 145.16 SMILES: CCOC(=O)CC(=O)NC