accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COO

CAS RN 7152-03-6

IUPAC Name: cyclopropyl(4-methoxyphenyl)methanone
Synonyms: cyclopropyl(4-methoxyphenyl)methanone
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: YKZSVEVTRUSPOQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)C1CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results