accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7162-59-6

IUPAC Name: 2-(hexyloxy)benzaldehyde
Synonyms: 2-(hexyloxy)benzaldehyde
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: IFOIDROUJIGQAV-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=CC=C1C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results