CAS RN 71676-01-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Amisulpride N-Oxide, TRC
CAS: 71676-01-2 Molecular Formula: C17 H27 N3 O5 S Molecular Weight (g/mol): 385.48 Synonym: Benzamide, 4-amino-N-[(1-ethyl-1-oxido-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxy-,Benzamide, 4-amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxy-, N-oxide,Amisulpride Imp. F (EP) IUPAC Name: 4-amino-N-[(1-ethyl-1-oxidopyrrolidin-1-ium-2-yl)methyl]-5-ethylsulfonyl-2-methoxybenzamide SMILES: CC[N+]1([O-])CCCC1CNC(=O)c2cc(c(N)cc2OC)S(=O)(=O)CC