missing translation for 'onlineSavingsMsg'
Learn More

Amisulpride N-Oxide, TRC

Product Code. 30138744
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138744 25 mg 25mg
30010845 10 mg 10mg
30004511 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138744 Supplier Toronto Research Chemicals Supplier No. TRCA63326525MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71676-01-2
Molecular Formula C17 H27 N3 O5 S
Molecular Weight (g/mol) 385.48
Synonym Benzamide, 4-amino-N-[(1-ethyl-1-oxido-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxy-, Benzamide, 4-amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxy-, N-oxide, Amisulpride Imp. F (EP)
IUPAC Name 4-amino-N-[(1-ethyl-1-oxidopyrrolidin-1-ium-2-yl)methyl]-5-ethylsulfonyl-2-methoxybenzamide
SMILES CC[N+]1([O-])CCCC1CNC(=O)c2cc(c(N)cc2OC)S(=O)(=O)CC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H27N3O5S/c1-4-20(22)8-6-7-12(20)11-19-17(21)13-9-16(26(23,24)5-2)14(18)10-15(13)25-3/h9-10,12H,4-8,11,18H2,1-3H3,(H,19,21)
Formula Weight 385.1671
Chemical Name or Material Amisulpride N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.