accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHBrN

CAS RN 7169-97-3

IUPAC Name: N-(5-bromopyridin-2-yl)acetamide
Synonyms: N-(5-bromopyridin-2-yl)acetamide
Molecular Weight (g/mol): 215.05
Molecular Formula: C7H7BrN2O
InChi Key: MJFCOXATGBYERZ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(Br)C=N1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results