CAS RN 720-44-5
IUPAC Name:
(4-methoxyphenyl)(phenyl)methanol
Synonyms:
(4-methoxyphenyl)(phenyl)methanol
Molecular Weight (g/mol):
214.26
Molecular Formula:
C14H14O2
InChi Key:
BEGZWXVLBIZFKQ-UHFFFAOYNA-N
SMILES:
COC1=CC=C(C=C1)C(O)C1=CC=CC=C1
1
–
1
of
1
results
1
4-Methoxybenzhydrol, 98+%
CAS: 720-44-5 Molecular Formula: C14H14O2 Molecular Weight (g/mol): 214.264 MDL Number: MFCD00014398 InChI Key: BEGZWXVLBIZFKQ-UHFFFAOYSA-N Synonym: 4-methoxybenzhydrol,4-methoxyphenyl phenyl methanol,4-methoxybenzhydryl alcohol,benzenemethanol, 4-methoxy-.alpha.-phenyl,4-methoxyphenyl phenylmethan-1-ol,4-methoxy-phenyl-phenyl-methanol,mamp-oh resin,4-methoxybenzohydrol,acmc-209olf,p-methoxybenzhydryl alcohol PubChem CID: 95375 IUPAC Name: (4-methoxyphenyl)-phenylmethanol SMILES: COC1=CC=C(C=C1)C(C2=CC=CC=C2)O