accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CCH3NHSH2N

CAS RN 7204-48-0

IUPAC Name: tert-butylthiourea
Synonyms: tert-butylthiourea
Molecular Weight (g/mol): 132.23
Molecular Formula: C5H12N2S
InChi Key: RYOCWONLFFPYMN-UHFFFAOYSA-N
SMILES: CC(C)(C)NC(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results