accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONO

CAS RN 723-89-7

IUPAC Name: 1-phenyl-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 1-phenylindole-2,3-dione
Molecular Weight (g/mol): 223.23
Molecular Formula: C14H9NO2
InChi Key: UWCPWBIMRYXUOU-UHFFFAOYSA-N
SMILES: O=C1N(C2=CC=CC=C2C1=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results