accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3O

CAS RN 7250-94-4

IUPAC Name: 2-acetylphenyl acetate
Synonyms: 2-acetylphenyl acetate
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: WWEDAIJJBDFOOK-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1C(C)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results