CAS RN 7291-22-7
Pyridine-d5, for NMR, contains 1 v/v% TMS, 99 atom % D
Pyridine-d5, for NMR, 100.0 atom % D
CAS: 7291-22-7 Molecular Formula: C5H5N Molecular Weight (g/mol): 84.13 MDL Number: MFCD00044639 InChI Key: JUJWROOIHBZHMG-RALIUCGRSA-N Synonym: pyridine-d5,2h5 pyridine,c5d5n,2 h? pyridine,pyridine d5,pyridine,crude,light,pyridine, perdeutero,∼2∼h_5_ pyridine,de85c PubChem CID: 558519 IUPAC Name: 2,3,4,5,6-pentadeuteriopyridine SMILES: [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
Pyridine-d{5}, 100%(Isotopic)
CAS: 7291-22-7 Molecular Formula: C5H5N Molecular Weight (g/mol): 84.13 MDL Number: MFCD00044639 InChI Key: JUJWROOIHBZHMG-RALIUCGRSA-N Synonym: pyridine-d5,2h5 pyridine,c5d5n,2 h? pyridine,pyridine d5,pyridine,crude,light,pyridine, perdeutero,∼2∼h_5_ pyridine,de85c PubChem CID: 558519 IUPAC Name: 2,3,4,5,6-pentadeuteriopyridine SMILES: [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
Pyridine-d5, for NMR, with 0.03% TMS, 100.0 atom% D
CAS: 7291-22-7 Molecular Formula: C5H5N Molecular Weight (g/mol): 84.13 MDL Number: MFCD00044639 InChI Key: JUJWROOIHBZHMG-RALIUCGRSA-N Synonym: pyridine-d5,2h5 pyridine,c5d5n,2 h? pyridine,pyridine d5,pyridine,crude,light,pyridine, perdeutero,∼2∼h_5_ pyridine,de85c PubChem CID: 558519 IUPAC Name: 2,3,4,5,6-pentadeuteriopyridine SMILES: [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
Pyridine-d{5}, 99.5% (Isotopic), contains 0.05% v/v TMS
CAS: 7291-22-7 Molecular Formula: C5H5N Molecular Weight (g/mol): 84.13 MDL Number: MFCD00044639 InChI Key: JUJWROOIHBZHMG-RALIUCGRSA-N Synonym: pyridine-d5,2h5 pyridine,c5d5n,2 h? pyridine,pyridine d5,pyridine,crude,light,pyridine, perdeutero,∼2∼h_5_ pyridine,de85c PubChem CID: 558519 IUPAC Name: 2,3,4,5,6-pentadeuteriopyridine SMILES: [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
Pyridine-d5, 99.5%(Isotopic), Thermo Scientific Chemicals
CAS: 7291-22-7 Molecular Formula: C5H5N Molecular Weight (g/mol): 84.13 MDL Number: MFCD00044639 InChI Key: JUJWROOIHBZHMG-RALIUCGRSA-N Synonym: pyridine-d5,2h5 pyridine,c5d5n,2 h? pyridine,pyridine d5,pyridine,crude,light,pyridine, perdeutero,∼2∼h_5_ pyridine,de85c PubChem CID: 558519 IUPAC Name: (²H₅)pyridine SMILES: [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
Pyridine-d5, for NMR, 99 atom % D
CAS: 7291-22-7 Molecular Formula: C5H5N Molecular Weight (g/mol): 84.13 MDL Number: MFCD00044639 InChI Key: JUJWROOIHBZHMG-RALIUCGRSA-N Synonym: pyridine-d5,2h5 pyridine,c5d5n,2 h? pyridine,pyridine d5,pyridine,crude,light,pyridine, perdeutero,∼2∼h_5_ pyridine,de85c PubChem CID: 558519 IUPAC Name: 2,3,4,5,6-pentadeuteriopyridine SMILES: [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
Pyridine-d{5}, 99.5% (Isotopic)
CAS: 7291-22-7 Molecular Formula: C5H5N Molecular Weight (g/mol): 84.13 MDL Number: MFCD00044639 InChI Key: JUJWROOIHBZHMG-RALIUCGRSA-N Synonym: pyridine-d5,2h5 pyridine,c5d5n,2 h? pyridine,pyridine d5,pyridine,crude,light,pyridine, perdeutero,∼2∼h_5_ pyridine,de85c PubChem CID: 558519 IUPAC Name: 2,3,4,5,6-pentadeuteriopyridine SMILES: [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D
Pyridine-d5 (Reagent grade), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
Pyridine-d5, TRC
CAS: 7291-22-7 Molecular Formula: C5 2H5 N Molecular Weight (g/mol): 84.13 Synonym: Pyridine-2,3,4,5,6-d5,Pyridine D5 (6CI,7CI,8CI,9CI),Pentadeuteropyridine,Perdeuteriopyridine IUPAC Name: 2,3,4,5,6-pentadeuteriopyridine SMILES: [2H]c1nc([2H])c([2H])c([2H])c1[2H]