accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

GeH4NHCH3H3CNHCH3CH3NHCH3H3CNHCH3H3C

CAS RN 7344-40-3

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 257
Molecular Formula: C8H32GeN4
InChi Key: RUPHDGAMPRMSPC-UHFFFAOYSA-N
SMILES: [GeH4].CNC.CNC.CNC.CNC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results