accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NClO

CAS RN 73568-25-9

IUPAC Name: 2-chloroquinoline-3-carbaldehyde
Synonyms: 2-chloroquinoline-3-carbaldehyde
Molecular Weight (g/mol): 191.61
Molecular Formula: C10H6ClNO
InChi Key: SDKQWXCBSNMYBN-UHFFFAOYSA-N
SMILES: ClC1=NC2=CC=CC=C2C=C1C=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)