CAS RN 738-70-5
Thermo Scientific Chemicals Trimethoprim
CAS: 738-70-5 Molecular Formula: C14H18N4O3 Synonym: 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine
Trimethoprim, 50 mg/ml in DMSO, sterile-filtered, Thermo Scientific Chemicals
CAS: 738-70-5 Molecular Formula: C14H18N4O3 Molecular Weight (g/mol): 290.323 MDL Number: MFCD00036761 InChI Key: IEDVJHCEMCRBQM-UHFFFAOYSA-N PubChem CID: 5578 ChEBI: CHEBI:45924 IUPAC Name: 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine SMILES: COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
Thermo Scientific Chemicals Trimethoprim, 98%
CAS: 738-70-5 Molecular Formula: C14H18N4O3 Molecular Weight (g/mol): 290.32 InChI Key: IEDVJHCEMCRBQM-UHFFFAOYSA-N Synonym: trimethoprim,proloprim,trimpex,bactramin,monotrim,monotrimin,trimetoprim,trimopan,monoprim,syraprim PubChem CID: 5578 ChEBI: CHEBI:45924 IUPAC Name: 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine SMILES: COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
Trimethoprim, 99.8%, MP Biomedicals™
CAS: 738-70-5 Molecular Formula: C14H18N4O3 Molecular Weight (g/mol): 290.323 InChI Key: IEDVJHCEMCRBQM-UHFFFAOYSA-N Synonym: 2,4-Diamino-5-[3,4,5-trimethoxy-benzyl]pyrimidine PubChem CID: 5578 ChEBI: CHEBI:45924 IUPAC Name: 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine SMILES: COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
Melford Trimethoprim
Trimethoprim is a bacteriostatic antibiotic that inhibits bacterial dihydrofolate reductase, an enzyme involved in the synthesis of tetrahydrofolic acid.
Trimethoprim, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Trimethoprim 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Trimethoprim, TRC
CAS: 738-70-5 Molecular Formula: C14 H18 N4 O3 Molecular Weight (g/mol): 290.32 Synonym: Trimethoprim,5-(3,4,5-Trimethoxybenzyl)pyrimidine-2,4-diamine,Pyrimidine, 2,4-diamino-5-(3,4,5-trimethoxybenzyl)- (7CI,8CI),2,4-Pyrimidinediamine, 5-[(3,4,5-trimethoxyphenyl)methyl]-,2,4-Diamino-5-(3',4',5'-trimethoxybenzyl)pyrimidine,2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine,BW 56-72,Instalac,Monotrim,NIH 204,NSC 106568,Proloprim,Syraprim,Tiempe,Trimanyl,Trimethioprim,Trimetoprim,Trimogal,Trimopan,Trimpex,Triprim,Uretrim,Wellcoprim IUPAC Name: 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine SMILES: COc1cc(Cc2cnc(N)nc2N)cc(OC)c1OC
Trimethoprim, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Trimethoprim, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TOKU-E™ Trimethoprim ReadyMade™ Solution
Trimethoprim ReadyMade™ Solution is provided as a sterile-filtered solution of Trimethoprim in DMSO.