missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Trimethoprim ReadyMade™ Solution

Product Code. 30044061
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044061 1 mL 1mL
30044016 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30044061 Supplier TOKU-E™ Supplier No. T1441ML
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Trimethoprim ReadyMade™ Solution is provided as a sterile-filtered solution of Trimethoprim in DMSO.

  • CAS Number: 738-70-5
  • Molecular Formula: C14H18N4O3
  • Molecular Weight: 290.32 g/mol
  • Storage Conditions: -20°C, Protect from light
  • Form: Liquid
  • Source: Synthetic
  • Spectrum: Trimethoprim has a wide antibacterial spectrum and is active against most Gram-positive and Gram-negative aerobic bacteria, including Nocardia, Brucella, Gram-negative bacilli, and some Gram-positive bacteria like Streptococcus
  • Mechanism of Action: Trimethoprim inhibits bacterial dihydrofolate reductase (DHFR) thus interfering with the conversion of dihydrofolate (DHF) to tetrahydrofolate (THF). THF is a precursor in the thymidine synthesis pathway and a disruption in this pathway inhibits bacterial the synthesis of bacterial DNA

Specifications

Chemical Name or Material Trimethoprim
Physical Form Liquid
CAS 738-70-5
Source Synthetic
Packaging 2 mL plastic vial
Sterile Filtered Yes, 0.22 μm
Sterility Sterile
Color Clear Colorless to Pale Yellow
Concentration 25.0 to 26.5 mg/mL
Quantity 1 mL
Flash Point 87°C
Purity Grade Notes >98% (HPLC)
Assay Percent Range 98.5 to 101.0%
Assay Percent Notes Trimethoprim, Dried Basis
Molecular Formula C14H18N4O3
Product Line ReadyMade™ Solutions
Shipping Condition Ice Pack
Synonym TMP, Proloprim, Trimpex
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
Molecular Weight (g/mol) 290.32 g/mol
Formula Weight 290.32 amu
Percent Purity >98%
Grade Antibiotic
Concentration or Composition (by Analyte or Components) Trimethoprim: ≥1 to <5%, DMSO ≥90 to ≤100%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.