accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ON

CAS RN 7391-28-8

IUPAC Name: 3-(4-methylphenyl)-3-oxopropanenitrile
Synonyms: 3-(4-methylphenyl)-3-oxopropanenitrile
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: AIECDYDQPCANJK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)CC#N
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)