accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFOO

CAS RN 73960-07-3

IUPAC Name: 4-(difluoromethoxy)benzaldehyde
Synonyms: 4-(difluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: ZWCXOJYJJINQGU-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results