CAS RN 7440-70-2
Calcium, 99%, pieces, Thermo Scientific Chemicals
CAS: 7440-70-2 Molecular Formula: Ca Molecular Weight (g/mol): 40.08 MDL Number: MFCD00010897 MFCD00085314 InChI Key: OYPRJOBELJOOCE-UHFFFAOYSA-N Synonym: unii-sy7q814vup,hsdb 273,sy7q814vup,compounds,elemental,calcium, elemental,kalzium,atom,ingot,chelate PubChem CID: 5460341 ChEBI: CHEBI:29320 IUPAC Name: calcium SMILES: [Ca]
Calcium standard solution, for AAS, 1 mg/ml Ca in 2 to 5% HNO3
CAS: 7440-70-2 | Ca | 40.08 g/mol
Calcium granules, redistilled, -6 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-70-2 Molecular Formula: Ca Molecular Weight (g/mol): 40.08 MDL Number: MFCD00010897 MFCD00085314 InChI Key: OYPRJOBELJOOCE-UHFFFAOYSA-N Synonym: unii-sy7q814vup,hsdb 273,sy7q814vup,compounds,elemental,calcium, elemental,kalzium,atom,ingot,chelate PubChem CID: 5460341 ChEBI: CHEBI:29320 IUPAC Name: calcium SMILES: [Ca]
Calcium turnings, 98% (metals basis), Thermo Scientific Chemicals
CAS: 7440-70-2 Molecular Formula: Ca Molecular Weight (g/mol): 40.08 MDL Number: MFCD00010897 MFCD00085314 InChI Key: OYPRJOBELJOOCE-UHFFFAOYSA-N Synonym: unii-sy7q814vup,hsdb 273,sy7q814vup,compounds,elemental,calcium, elemental,kalzium,atom,ingot,chelate PubChem CID: 5460341 ChEBI: CHEBI:29320 IUPAC Name: calcium SMILES: [Ca]
Calcium crystalline dendritic pieces, 99.98% (metals basis), Thermo Scientific Chemicals
CAS: 7440-70-2 Molecular Formula: Ca Molecular Weight (g/mol): 40.08 MDL Number: MFCD00010897 MFCD00085314 InChI Key: OYPRJOBELJOOCE-UHFFFAOYSA-N Synonym: unii-sy7q814vup,hsdb 273,sy7q814vup,compounds,elemental,calcium, elemental,kalzium,atom,ingot,chelate PubChem CID: 5460341 ChEBI: CHEBI:29320 IUPAC Name: calcium SMILES: [Ca]
Calcium granules, redistilled, -16 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-70-2 Molecular Formula: Ca Molecular Weight (g/mol): 40.08 MDL Number: MFCD00010897 MFCD00085314 InChI Key: OYPRJOBELJOOCE-UHFFFAOYSA-N Synonym: unii-sy7q814vup,hsdb 273,sy7q814vup,compounds,elemental,calcium, elemental,kalzium,atom,ingot,chelate PubChem CID: 5460341 ChEBI: CHEBI:29320 IUPAC Name: calcium SMILES: [Ca]
Calcium granules, -9 mesh, 98.8% (metals basis), Thermo Scientific Chemicals
CAS: 7440-70-2 Molecular Formula: Ca Molecular Weight (g/mol): 40.08 MDL Number: MFCD00010897 MFCD00085314 InChI Key: OYPRJOBELJOOCE-UHFFFAOYSA-N Synonym: unii-sy7q814vup,hsdb 273,sy7q814vup,compounds,elemental,calcium, elemental,kalzium,atom,ingot,chelate PubChem CID: 5460341 ChEBI: CHEBI:29320 IUPAC Name: calcium SMILES: [Ca]
Calcium Single Element Ion Standard - Cation, SPEX CertiPrep™
Calcium Standard: Ca+2 @ 1000 μg/mL in H2O, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Standard 1607: Ca @ 2000; Mo, P, Zn @ 800 μg/g in 75 cSt Oil
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Water Supply QC Check Sample: Hardness
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Sulfur-Free Standard 1604: Ca @ 5000; Mg, Mo, P, Zn @ 1600 μg/g in 75 cSt Oil
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lubricating Oil Standards Set (contains one of each LOIL1 thru LOIL16), VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications