accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 74457-86-6

H3COCH3OF

CAS RN 74457-86-6

IUPAC Name: 1-(2-fluoro-4-methoxyphenyl)ethan-1-one
Synonyms: 1-(2-fluoro-4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: PIRRWUMTIBFCCW-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(C)=O)C(F)=C1
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)